siRNA Details
| Gene Symbol: Stra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029924.1 | NP_001025095.1 | 6 | 542 | ||
| GenBank mRNA | BC097373.1 | - | 6 | 542 | 7444462 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stra6| Gene Symbol: Stra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029924.1 | NP_001025095.1 | 6 | 542 | ||
| GenBank mRNA | BC097373.1 | - | 6 | 542 | 7444462 | |