siRNA Details
| Gene Symbol: Megf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243281.2 | XP_006243343.1 | 24 | 3539 | ||
| XM_008766385.1 | XP_008764607.1 | 23 | 3458 | |||
| XM_008766389.1 | XP_008764611.1 | 21 | 3539 | |||
| XM_008766390.1 | XP_008764612.1 | 20 | 3014 | |||
| XM_008766392.1 | XP_008764614.1 | 20 | 3035 | |||

Megf11
Genomic Map