siRNA Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009470.1 | NP_001009470.1 | 6 | 793 | ||
| GenBank mRNA | BC088212.1 | - | 6 | 793 | 7383459 | |
| BC097952.1 | - | 6 | 817 | 7454811 | ||
| FQ234119.1 | - | 6 | 807 | |||
| Gene Symbol: LOC100364016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758651.1 | XP_008756873.1 | 1 | 827 | ||

Ccnb2
Genomic Map