siRNA Details
| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014211.1 | NP_001014233.1 | 6 | 569 | ||
| XM_008766312.1 | XP_008764534.1 | 7 | 755 | |||
| GenBank mRNA | BC089899.1 | - | 6 | 569 | 7382871 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myzap| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014211.1 | NP_001014233.1 | 6 | 569 | ||
| XM_008766312.1 | XP_008764534.1 | 7 | 755 | |||
| GenBank mRNA | BC089899.1 | - | 6 | 569 | 7382871 | |