siRNA Details
Gene Symbol: Mns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007752.1 | NP_001007753.1 | 4 | 555 | ||
GenBank mRNA | BC079390.1 | - | 4 | 555 | 7111791 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007752.1 | NP_001007753.1 | 4 | 555 | ||
GenBank mRNA | BC079390.1 | - | 4 | 555 | 7111791 |