siRNA Details
| Gene Symbol: Rwdd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108773.1 | NP_001102243.1 | 2, 3 | 477 | ||
| XM_006243483.2 | XP_006243545.1 | 2, 3 | 364 | |||
| XM_006243485.2 | XP_006243547.1 | 2, 3 | 403 | |||
| XM_006243486.2 | XP_006243548.1 | 2, 3 | 580 | |||

Rwdd2a
Genomic Map