siRNA Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079855.1 | NP_001073324.1 | 6 | 783 | ||
| NM_001184702.1 | NP_001171631.1 | 6 | 783 | |||
| NM_001184703.1 | NP_001171632.1 | 7 | 876 | |||
| NM_001184704.1 | NP_001171633.1 | 5 | 641 | |||
| NM_003918.2 | NP_003909.2 | 7 | 876 | |||
| XM_011545600.2 | XP_011543902.1 | 6 | 714 | |||
| XM_017029927.1 | XP_016885416.1 | 7 | 826 | |||
| XM_017029928.1 | XP_016885417.1 | 7 | 801 | |||
| XM_017029929.1 | XP_016885418.1 | 7 | 986 | |||
| XM_017029930.1 | XP_016885419.1 | 6 | 691 | |||
| XM_017029931.1 | XP_016885420.1 | 6 | 714 | |||
| GenBank mRNA | AK098046.1 | - | 5 | 549 | ||
| AK292496.1 | - | 6 | 691 | |||
| BC023152.1 | - | 6 | 650 | 3923041 | ||
| DQ424901.1 | - | 5 | 594 | |||
| U94358.1 | - | 3 | 272 | |||
| U94360.1 | - | 3 | 331 | |||
| U94361.1 | - | 2 | 49 | |||
| U94362.1 | - | 7 | 877 | |||
| U94363.1 | - | 6 | 657 | |||

GYG2
Genomic Map