siRNA Details
| Gene Symbol: Dnajc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750579.2 | XP_003750627.1 | 6 | 720 | ||
| XM_008766592.1 | XP_008764814.1 | 5 | 484 | |||
| XM_008766593.1 | XP_008764815.1 | 6 | 720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc13| Gene Symbol: Dnajc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750579.2 | XP_003750627.1 | 6 | 720 | ||
| XM_008766592.1 | XP_008764814.1 | 5 | 484 | |||
| XM_008766593.1 | XP_008764815.1 | 6 | 720 | |||