siRNA Details
| Gene Symbol: Aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040156.2 | NP_001035246.2 | 4, 5 | 1965 | ||
| XM_006243672.2 | XP_006243734.1 | 3, 4 | 1720 | |||
| XM_006243675.2 | XP_006243737.1 | 4, 5 | 1612 | |||
| XM_008766543.1 | XP_008764765.1 | 3, 4 | 2149 | |||
| XM_008766544.1 | XP_008764766.1 | 3, 4 | 2149 | |||
| XM_008766545.1 | XP_008764767.1 | 3, 4 | 844 | |||
| XM_008766546.1 | XP_008764768.1 | 3, 4 | 490 | |||
| XM_008766547.1 | XP_008764769.1 | 2, 3 | 340 | |||
| XM_008766548.1 | XP_008764770.1 | 3, 4 | 384 | |||
| GenBank mRNA | BC083913.1 | - | 4, 5 | 512 | ||

Aste1
Genomic Map