siRNA Details
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108777.1 | NP_001102247.1 | 4 | 1452 | ||
XM_006243800.1 | XP_006243862.1 | 4 | 1479 | |||
XM_006243801.2 | XP_006243863.1 | 4 | 1130 | |||
GenBank mRNA | BC168984.1 | - | 4 | 1424 | 7103631 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
