siRNA Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079942.1 | NP_001073411.1 | 14 | 1600 | ||
| XM_006243809.2 | XP_006243871.1 | 15 | 1682 | |||
| XM_006243810.2 | XP_006243872.1 | 14 | 2570 | |||
| XM_008766550.1 | XP_008764772.1 | 15 | 2258 | |||
| XM_008766551.1 | XP_008764773.1 | 13 | 1554 | |||
| GenBank mRNA | BC129103.1 | - | 14 | 1600 | 7100098 | |

Sema3b
Genomic Map