siRNA Details
| Gene Symbol: Gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108781.1 | NP_001102251.1 | 5 | 700 | ||
| XM_006243811.2 | XP_006243873.1 | 6 | 997 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmppb| Gene Symbol: Gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108781.1 | NP_001102251.1 | 5 | 700 | ||
| XM_006243811.2 | XP_006243873.1 | 6 | 997 | |||