siRNA Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014214.1 | NP_001014236.1 | 11 | 1473 | ||
| GenBank mRNA | BC082046.1 | - | 11 | 1473 | 7131735 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dalrd3| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014214.1 | NP_001014236.1 | 11 | 1473 | ||
| GenBank mRNA | BC082046.1 | - | 11 | 1473 | 7131735 | |