siRNA Details
| Gene Symbol: Ccdc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108783.1 | NP_001102253.1 | 5, 6 | 402 | ||
| XM_006243970.2 | XP_006244032.1 | 6, 7 | 1026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc12| Gene Symbol: Ccdc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108783.1 | NP_001102253.1 | 5, 6 | 402 | ||
| XM_006243970.2 | XP_006244032.1 | 6, 7 | 1026 | |||