siRNA Details
| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135259.2 | NP_001128731.1 | 3 | 305 | ||
| NM_001199225.1 | NP_001186154.1 | 3 | 305 | |||
| XM_006244034.2 | XP_006244096.1 | 4 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmc1| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135259.2 | NP_001128731.1 | 3 | 305 | ||
| NM_001199225.1 | NP_001186154.1 | 3 | 305 | |||
| XM_006244034.2 | XP_006244096.1 | 4 | 371 | |||