siRNA Details
| Gene Symbol: Lyzl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001246183.1 | NP_001233112.1 | 2, 3 | 295 | ||
| XM_006244123.2 | XP_006244185.2 | 3, 4 | 1538 | |||
| XM_006244125.2 | XP_006244187.1 | 3, 4 | 491 | |||
| GenBank mRNA | HM125534.1 | - | 2, 3 | 295 | ||

Lyzl4
Genomic Map