siRNA Details
| Gene Symbol: Tcaim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110838.1 | NP_001104308.1 | 6 | 795 | ||
| XM_006244192.2 | XP_006244254.1 | 6 | 907 | |||
| XM_006244193.2 | XP_006244255.1 | 6 | 1016 | |||
| XM_006244194.2 | XP_006244256.1 | 6 | 932 | |||
| XM_006244195.2 | XP_006244257.1 | 6 | 900 | |||
| XM_008766717.1 | XP_008764939.1 | 6 | 932 | |||
| XM_008766718.1 | XP_008764940.1 | 5 | 1041 | |||
| XM_008766719.1 | XP_008764941.1 | 5 | 799 | |||
| XM_008766720.1 | XP_008764942.1 | 5 | 764 | |||
| XM_008766721.1 | XP_008764943.1 | 5 | 796 | |||
| XM_008766722.1 | XP_008764944.1 | 6 | 906 | |||
| XM_008766723.1 | XP_008764945.1 | 5 | 911 | |||
| XM_008766724.1 | XP_008764946.1 | 6 | 824 | |||
| XM_008766725.1 | XP_008764947.1 | 6 | 828 | |||
| XM_008766726.1 | XP_008764948.1 | 5 | 692 | |||
| GenBank mRNA | FQ210480.1 | - | 6 | 809 | ||
| FQ211645.1 | - | 6 | 880 | |||
| FQ215070.1 | - | 6 | 809 | |||

Tcaim
Genomic Map