siRNA Details
| Gene Symbol: CACNA1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005407.1 | NP_001005407.1 | 9 | 2214 | ||
| NM_021098.2 | NP_066921.2 | 9 | 2214 | |||
| XM_005255652.4 | XP_005255709.1 | 9 | 2060 | |||
| XM_006720963.3 | XP_006721026.1 | 9 | 2060 | |||
| XM_006720964.3 | XP_006721027.1 | 9 | 2060 | |||
| XM_006720965.3 | XP_006721028.1 | 9 | 2060 | |||
| XM_006720967.3 | XP_006721030.1 | 9 | 2061 | |||
| XM_006720968.3 | XP_006721031.1 | 9 | 2061 | |||
| XM_011522724.2 | XP_011521026.1 | 6 | 1509 | |||
| XM_011522727.2 | XP_011521029.1 | 9 | 2067 | |||
| XM_017023819.1 | XP_016879308.1 | 9 | 2021 | |||
| XM_017023820.1 | XP_016879309.1 | 9 | 2060 | |||
| XM_017023821.1 | XP_016879310.1 | 9 | 2060 | |||
| GenBank mRNA | AF051946.3 | - | 9 | 2045 | ||
| AF073931.1 | - | 9 | 2214 | |||
| AJ420779.1 | - | 8 | 1966 | |||
| BM554380.1 | - | 1 | 695 | |||

CACNA1H
Genomic Map