siRNA Details
| Gene Symbol: Mrps10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008859.1 | NP_001008859.1 | 3 | 134 | ||
| XM_006244464.2 | XP_006244526.1 | 4 | 283 | |||
| XM_006244465.2 | XP_006244527.1 | 4 | 280 | |||
| GenBank mRNA | AY310148.1 | - | 3 | 134 | ||

Mrps10
Genomic Map