siRNA Details
| Gene Symbol: Ubr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178071.1 | NP_001171542.1 | 15 | 1786 | ||
| GenBank mRNA | AB549212.1 | - | 15 | 1786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr2| Gene Symbol: Ubr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178071.1 | NP_001171542.1 | 15 | 1786 | ||
| GenBank mRNA | AB549212.1 | - | 15 | 1786 | ||