siRNA Details
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001062510.5 | XP_001062510.4 | 8 | 1509 | ||
XM_006244570.2 | XP_006244632.1 | 8 | 1512 | |||
GenBank mRNA | BC099800.1 | - | 7 | 801 |
Gene Symbol: LOC100909464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750611.3 | XP_003750659.3 | 5 | 643 |
