siRNA Details
| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750650.3 | XP_003750698.1 | 20 | 1996 | ||
| XM_006244582.2 | XP_006244644.1 | 19 | 1958 | |||
| XM_006244583.1 | XP_006244645.1 | 19 | 2038 | |||
| XM_008766903.1 | XP_008765125.1 | 20 | 1986 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_198167, BC059283, BC039277, BC026370, BC057136, and others.

Tmem63b
Genomic Map