siRNA Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014215.1 | NP_001014237.1 | 4 | 442 | ||
| GenBank mRNA | BC082113.1 | - | 4 | 442 | 7135459 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpq| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014215.1 | NP_001014237.1 | 4 | 442 | ||
| GenBank mRNA | BC082113.1 | - | 4 | 442 | 7135459 | |