siRNA Details
| Gene Symbol: Phf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108791.1 | NP_001102261.1 | 4 | 766 | ||
| XM_006244673.2 | XP_006244735.1 | 3 | 452 | |||
| XM_006244675.2 | XP_006244737.1 | 3 | 619 | |||
| XM_006244676.2 | XP_006244738.1 | 3 | 512 | |||
| XM_006244677.2 | XP_006244739.1 | 4 | 436 | |||
| XM_008766954.1 | XP_008765176.1 | 4 | 389 | |||

Phf3
Genomic Map