siRNA Details
| Gene Symbol: Ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108793.1 | NP_001102263.1 | 4 | 838 | ||
| GenBank mRNA | BC166583.1 | - | 403 | 7442489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc115| Gene Symbol: Ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108793.1 | NP_001102263.1 | 4 | 838 | ||
| GenBank mRNA | BC166583.1 | - | 403 | 7442489 | ||