siRNA Details
| Gene Symbol: Mitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009714.1 | NP_001009714.1 | 588 | |||
| XM_006244828.2 | XP_006244890.1 | 626 | ||||
| XM_008767036.1 | XP_008765258.1 | 662 | ||||
| GenBank mRNA | BC088263.1 | - | 588 | 7376620 | ||

Mitd1
Genomic Map