siRNA Details
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191887.1 | NP_001178816.1 | 17 | 3038 | ||
| XM_008767037.1 | XP_008765259.1 | 13 | 4567 | |||
| XM_008767038.1 | XP_008765260.1 | 13 | 4564 | |||
| XM_008767039.1 | XP_008765261.1 | 13 | 4567 | |||

Aff3
Genomic Map