siRNA Details
| Gene Symbol: Cnot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108794.2 | NP_001102264.1 | 7 | 1428 | ||
| GenBank mRNA | BC158750.1 | - | 7 | 1428 | 9101674 | |
| FQ233266.1 | - | 7 | 1433 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnot11| Gene Symbol: Cnot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108794.2 | NP_001102264.1 | 7 | 1428 | ||
| GenBank mRNA | BC158750.1 | - | 7 | 1428 | 9101674 | |
| FQ233266.1 | - | 7 | 1433 | |||