siRNA Details
| Gene Symbol: Nabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014216.1 | NP_001014238.1 | 2, 3 | 425 | ||
| GenBank mRNA | BC089852.1 | - | 2, 3 | 425 | 7377638 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nabp1| Gene Symbol: Nabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014216.1 | NP_001014238.1 | 2, 3 | 425 | ||
| GenBank mRNA | BC089852.1 | - | 2, 3 | 425 | 7377638 | |