siRNA Details
| Gene Symbol: Tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108795.1 | NP_001102265.1 | 3 | 801 | ||
| XM_008767117.1 | XP_008765339.1 | 3 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmeff2| Gene Symbol: Tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108795.1 | NP_001102265.1 | 3 | 801 | ||
| XM_008767117.1 | XP_008765339.1 | 3 | 796 | |||