siRNA Details
| Gene Symbol: Tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108795.1 | NP_001102265.1 | 6, 7 | 1108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmeff2| Gene Symbol: Tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108795.1 | NP_001102265.1 | 6, 7 | 1108 | ||