siRNA Details
| Gene Symbol: Bzw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198789.2 | NP_942084.1 | 6 | 526 | ||
| XM_006245000.2 | XP_006245062.2 | 6 | 525 | |||
| GenBank mRNA | BC061580.1 | - | 6 | 530 | 6890414 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bzw1| Gene Symbol: Bzw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198789.2 | NP_942084.1 | 6 | 526 | ||
| XM_006245000.2 | XP_006245062.2 | 6 | 525 | |||
| GenBank mRNA | BC061580.1 | - | 6 | 530 | 6890414 | |