siRNA Details
| Gene Symbol: Als2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013413.1 | NP_001013431.1 | 9 | 2069 | ||
| XM_006245001.2 | XP_006245063.1 | 10 | 2404 | |||
| XM_008767118.1 | XP_008765340.1 | 10 | 2410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2| Gene Symbol: Als2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013413.1 | NP_001013431.1 | 9 | 2069 | ||
| XM_006245001.2 | XP_006245063.1 | 10 | 2404 | |||
| XM_008767118.1 | XP_008765340.1 | 10 | 2410 | |||