siRNA Details
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177419.2 | NP_803154.1 | 11 | 1286 | ||
GenBank mRNA | AB066103.1 | - | 9 | 1043 | ||
BC061576.1 | - | 11 | 1289 | 6887693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177419.2 | NP_803154.1 | 11 | 1286 | ||
GenBank mRNA | AB066103.1 | - | 9 | 1043 | ||
BC061576.1 | - | 11 | 1289 | 6887693 |