siRNA Details
| Gene Symbol: Nhej1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014217.1 | NP_001014239.1 | 4 | 561 | ||
| XM_006245240.1 | XP_006245302.1 | 4 | 639 | |||
| XM_006245241.2 | XP_006245303.1 | 4 | 1215 | |||
| XM_006245242.2 | XP_006245304.1 | 4 | 984 | |||
| GenBank mRNA | BC079033.1 | - | 4 | 561 | 7114269 | |

Nhej1
Genomic Map