siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100760.1 | NP_001094230.1 | 6 | 829 | ||
| XM_006245244.2 | XP_006245306.1 | 6 | 851 | |||
| GenBank mRNA | BC104673.1 | - | 6 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam134a| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100760.1 | NP_001094230.1 | 6 | 829 | ||
| XM_006245244.2 | XP_006245306.1 | 6 | 851 | |||
| GenBank mRNA | BC104673.1 | - | 6 | 639 | ||