siRNA Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014218.1 | NP_001014240.1 | 5 | 731 | ||
| XM_006245247.2 | XP_006245309.1 | 5 | 435 | |||
| XM_008767262.1 | XP_008765484.1 | 5 | 1098 | |||
| XM_008767263.1 | XP_008765485.1 | 5 | 610 | |||
| XM_008767264.1 | XP_008765486.1 | 5 | 438 | |||
| XM_008767265.1 | XP_008765487.1 | 6 | 496 | |||
| XM_008767266.1 | XP_008765488.1 | 6 | 499 | |||
| GenBank mRNA | BC089204.1 | - | 5 | 731 | 7190326 | |

Atg9a
Genomic Map