siRNA Details
| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108806.1 | NP_001102276.1 | 3 | 368 | ||
| XM_006245413.2 | XP_006245475.1 | 3 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde6d| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108806.1 | NP_001102276.1 | 3 | 368 | ||
| XM_006245413.2 | XP_006245475.1 | 3 | 416 | |||