siRNA Details
| Gene Symbol: Prss56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750730.3 | XP_003750778.1 | 10 | 1379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss56| Gene Symbol: Prss56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750730.3 | XP_003750778.1 | 10 | 1379 | ||