siRNA Details
| Gene Symbol: Atg16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108809.3 | NP_001102279.2 | 2 | 180 | ||
| XM_006245419.2 | XP_006245481.1 | 2 | 358 | |||
| XM_006245420.2 | XP_006245482.1 | 2 | 357 | |||
| GenBank mRNA | CB605957.1 | - | 2 | 202 | ||

Atg16l1
Genomic Map