siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013227.2 | NP_001013245.1 | 2, 3 | 199 | ||
| GenBank mRNA | BC063182.1 | - | 2, 3 | 203 | 6887869 | |
| FQ215552.1 | - | 2, 3 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cops8| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013227.2 | NP_001013245.1 | 2, 3 | 199 | ||
| GenBank mRNA | BC063182.1 | - | 2, 3 | 203 | 6887869 | |
| FQ215552.1 | - | 2, 3 | 198 | |||