siRNA Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007755.1 | NP_001007756.1 | 3, 4 | 286 | ||
| XM_006245431.2 | XP_006245493.1 | 3, 4 | 862 | |||
| XM_008767276.1 | XP_008765498.1 | 3, 4 | 890 | |||
| XM_008767277.1 | XP_008765499.1 | 3, 4 | 365 | |||
| GenBank mRNA | BC079358.1 | - | 3, 4 | 286 | 7108615 | |

Scly
Genomic Map