siRNA Details
| Gene Symbol: Hdac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053449.1 | NP_445901.1 | 14 | 2099 | ||
| XM_006245438.1 | XP_006245500.1 | 14 | 2326 | |||
| XM_006245440.1 | XP_006245502.1 | 14 | 2311 | |||
| XM_006245441.1 | XP_006245503.1 | 14 | 2326 | |||
| XM_006245442.1 | XP_006245504.1 | 14 | 2311 | |||
| XM_006245443.1 | XP_006245505.1 | 14 | 2103 | |||
| XM_008767278.1 | XP_008765500.1 | 13 | 2095 | |||
| XM_008767279.1 | XP_008765501.1 | 13 | 7986 | |||

Hdac4
Genomic Map