siRNA Details
| Gene Symbol: BSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003458.3 | NP_003449.2 | 5 | 3402 | ||
| GenBank mRNA | AF052224.1 | - | 5 | 3403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BSN| Gene Symbol: BSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003458.3 | NP_003449.2 | 5 | 3402 | ||
| GenBank mRNA | AF052224.1 | - | 5 | 3403 | ||