siRNA Details
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126289.1 | NP_001119761.1 | 12 | 1675 | ||
GenBank mRNA | BC160823.1 | - | 12 | 1675 | 7380256 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126289.1 | NP_001119761.1 | 12 | 1675 | ||
GenBank mRNA | BC160823.1 | - | 12 | 1675 | 7380256 |