siRNA Details
| Gene Symbol: Fundc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025027.1 | NP_001020198.1 | 2 | 180 | ||
| GenBank mRNA | BC091353.1 | - | 2 | 180 | 7317238 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fundc1| Gene Symbol: Fundc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025027.1 | NP_001020198.1 | 2 | 180 | ||
| GenBank mRNA | BC091353.1 | - | 2 | 180 | 7317238 | |