siRNA Details
| Gene Symbol: PHOX2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003924.3 | NP_003915.2 | 3 | 1248 | ||
| GenBank mRNA | BC017199.2 | - | 3 | 984 | 3161284 | |
| D82344.1 | - | 3 | 1248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHOX2B| Gene Symbol: PHOX2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003924.3 | NP_003915.2 | 3 | 1248 | ||
| GenBank mRNA | BC017199.2 | - | 3 | 984 | 3161284 | |
| D82344.1 | - | 3 | 1248 | |||