siRNA Details
| Gene Symbol: Dynlt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013228.1 | NP_001013246.1 | 3 | 135 | ||
| GenBank mRNA | BC083797.1 | - | 3 | 135 | 7108507 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dynlt3| Gene Symbol: Dynlt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013228.1 | NP_001013246.1 | 3 | 135 | ||
| GenBank mRNA | BC083797.1 | - | 3 | 135 | 7108507 | |