siRNA Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033899.1 | NP_001029071.1 | 5 | 473 | ||
| GenBank mRNA | BC101870.1 | - | 5 | 473 | 5621864 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sms| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033899.1 | NP_001029071.1 | 5 | 473 | ||
| GenBank mRNA | BC101870.1 | - | 5 | 473 | 5621864 | |