siRNA Details
| Gene Symbol: RGD1566265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134589.1 | NP_001128061.1 | 2 | 211 | ||
| GenBank mRNA | FQ229129.1 | - | 2 | 225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566265| Gene Symbol: RGD1566265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134589.1 | NP_001128061.1 | 2 | 211 | ||
| GenBank mRNA | FQ229129.1 | - | 2 | 225 | ||